siRNA Details
| Gene Symbol: Terf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108448.1 | NP_001101918.1 | 7, 8 | 1255 | ||
| XM_006255564.2 | XP_006255626.1 | 9, 10 | 1670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Terf2| Gene Symbol: Terf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108448.1 | NP_001101918.1 | 7, 8 | 1255 | ||
| XM_006255564.2 | XP_006255626.1 | 9, 10 | 1670 | |||