siRNA Details
| Gene Symbol: NEWGENE_1305243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_341690.7 | XP_341691.7 | 160 | |||
| GenBank mRNA | FQ230829.1 | - | 112 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NEWGENE_1305243| Gene Symbol: NEWGENE_1305243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_341690.7 | XP_341691.7 | 160 | |||
| GenBank mRNA | FQ230829.1 | - | 112 | |||