siRNA Details
| Gene Symbol: Hsdl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024896.1 | NP_001020067.1 | 3 | 1035 | ||
| XM_006255712.2 | XP_006255774.1 | 4 | 1019 | |||
| XM_006255713.2 | XP_006255775.1 | 4 | 812 | |||
| XM_006255714.2 | XP_006255776.1 | 4 | 1972 | |||
| XM_008772622.1 | XP_008770844.1 | 5 | 1034 | |||
| GenBank mRNA | BC097457.1 | - | 3 | 1035 | 7457192 | |

Hsdl1
Genomic Map