siRNA Details
| Gene Symbol: Dnaaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014154.1 | NP_001014176.1 | 10 | 1439 | ||
| GenBank mRNA | BC079038.1 | - | 10 | 1439 | 7114363 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnaaf1| Gene Symbol: Dnaaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014154.1 | NP_001014176.1 | 10 | 1439 | ||
| GenBank mRNA | BC079038.1 | - | 10 | 1439 | 7114363 | |