siRNA Details
| Gene Symbol: Taf1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014155.1 | NP_001014177.1 | 11 | 1109 | ||
| XM_008772623.1 | XP_008770845.1 | 10 | 1289 | |||
| XM_008772624.1 | XP_008770846.1 | 9 | 1923 | |||
| XM_008772625.1 | XP_008770847.1 | 7 | 667 | |||
| GenBank mRNA | BC061804.1 | - | 11 | 1109 | 5598458 | |
| BC062077.1 | - | 6 | 1985 | |||

Taf1c
Genomic Map