siRNA Details
| Gene Symbol: Cotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108452.1 | NP_001101922.1 | 4 | 502 | ||
| GenBank mRNA | BC158746.1 | - | 4 | 481 | 9100871 | |
| EF076769.1 | - | 4 | 375 | |||
| FQ232981.1 | - | 4 | 478 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cotl1| Gene Symbol: Cotl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108452.1 | NP_001101922.1 | 4 | 502 | ||
| GenBank mRNA | BC158746.1 | - | 4 | 481 | 9100871 | |
| EF076769.1 | - | 4 | 375 | |||
| FQ232981.1 | - | 4 | 478 | |||