siRNA Details
| Gene Symbol: Fam92b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772654.1 | XP_008770876.1 | 3 | 434 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam92b| Gene Symbol: Fam92b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772654.1 | XP_008770876.1 | 3 | 434 | ||