siRNA Details
Gene Symbol: Emc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012165.1 | NP_001012165.1 | 2 | 368 | ||
GenBank mRNA | BC089914.1 | - | 2 | 368 | 7305045 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Emc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012165.1 | NP_001012165.1 | 2 | 368 | ||
GenBank mRNA | BC089914.1 | - | 2 | 368 | 7305045 |