siRNA Details
| Gene Symbol: Cbfa2t3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108453.1 | NP_001101923.1 | 9, 10 | 1423 | ||
| XM_006255770.2 | XP_006255832.1 | 8, 9 | 2317 | |||
| XM_006255772.2 | XP_006255834.1 | 9, 10 | 1578 | |||
| XM_008772629.1 | XP_008770851.1 | 8, 9 | 3288 | |||
| XM_008772630.1 | XP_008770852.1 | 7, 8 | 3211 | |||
| XM_008772631.1 | XP_008770853.1 | 8, 9 | 1773 | |||

Cbfa2t3
Genomic Map