siRNA Details
| Gene Symbol: Cdh15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207613.1 | NP_997496.1 | 12 | 1956 | ||
| GenBank mRNA | AB176538.1 | - | 12 | 1956 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh15| Gene Symbol: Cdh15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207613.1 | NP_997496.1 | 12 | 1956 | ||
| GenBank mRNA | AB176538.1 | - | 12 | 1956 | ||