siRNA Details
| Gene Symbol: Cdk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025722.2 | NP_001020893.2 | 6 | 967 | ||
| NM_001109936.1 | NP_001103406.1 | 7 | 516 | |||
| NM_001109937.1 | NP_001103407.1 | 7 | 477 | |||
| GenBank mRNA | BC098804.1 | - | 6 | 967 | 7376321 | |
| DN932869.1 | - | 7 | 512 | |||
| DN935585.1 | - | 7 | 536 | |||
| DQ607073.1 | - | 1 | 18 | |||
| DY320517.1 | - | 7 | 517 | |||

Cdk10
Genomic Map