siRNA Details
| Gene Symbol: Fanca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108455.1 | NP_001101925.1 | 6 | 601 | ||
| XM_008772633.1 | XP_008770855.1 | 6 | 612 | |||
| Gene Symbol: LOC103690042 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772669.1 | XP_008770891.1 | 6 | 614 | ||

Fanca
Genomic Map