siRNA Details
| Gene Symbol: Afg3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108456.3 | NP_001101926.3 | 8 | 1000 | ||
| GenBank mRNA | BC168848.1 | - | 8 | 893 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Afg3l1| Gene Symbol: Afg3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108456.3 | NP_001101926.3 | 8 | 1000 | ||
| GenBank mRNA | BC168848.1 | - | 8 | 893 | ||