siRNA Details
| Gene Symbol: Fam89a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011711.1 | NP_001011711.1 | 2 | 1304 | ||
| GenBank mRNA | AY569012.1 | - | 2 | 1304 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam89a| Gene Symbol: Fam89a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011711.1 | NP_001011711.1 | 2 | 1304 | ||
| GenBank mRNA | AY569012.1 | - | 2 | 1304 | ||