siRNA Details
| Gene Symbol: Ginm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227626.2 | XP_006227688.1 | 3, 4 | 327 | ||
| XM_006227627.2 | XP_006227689.1 | 3, 4 | 321 | |||
| XM_341726.7 | XP_341727.4 | 3, 4 | 328 | |||
| GenBank mRNA | FQ228838.1 | - | 277 | |||

Ginm1
Genomic Map