siRNA Details
| Gene Symbol: Ginm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227626.2 | XP_006227688.1 | 5 | 526 | ||
| XM_006227627.2 | XP_006227689.1 | 5 | 520 | |||
| XM_341726.7 | XP_341727.4 | 5 | 527 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ginm1| Gene Symbol: Ginm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006227626.2 | XP_006227688.1 | 5 | 526 | ||
| XM_006227627.2 | XP_006227689.1 | 5 | 520 | |||
| XM_341726.7 | XP_341727.4 | 5 | 527 | |||