siRNA Details
| Gene Symbol: Ccdc28a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037789.1 | NP_001032878.1 | 4, 5 | 627 | ||
| GenBank mRNA | BC100268.1 | - | 4, 5 | 627 | 7385321 | |
| FQ212615.1 | - | 4, 5 | 632 | |||
| FQ213153.1 | - | 4, 5 | 660 | |||

Ccdc28a
Genomic Map