siRNA Details
| Gene Symbol: Echdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007734.1 | NP_001007735.1 | 3 | 377 | ||
| XM_006227746.2 | XP_006227808.1 | 3 | 567 | |||
| XM_006227747.2 | XP_006227809.1 | 3 | 1164 | |||
| GenBank mRNA | BC079052.1 | - | 3 | 377 | 7115386 | |

Echdc1
Genomic Map