siRNA Details
| Gene Symbol: Rsph3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748694.3 | XP_003748742.1 | 1 | 1706 | ||
| XM_006227909.2 | XP_006227971.1 | 1 | 1706 | |||
| XM_006227910.2 | XP_006227972.1 | 1 | 1706 | |||
| XM_006227911.2 | XP_006227973.1 | 1 | 1706 | |||
| XM_008758741.1 | XP_008756963.1 | 1 | 1706 | |||
| XM_008758742.1 | XP_008756964.1 | 1 | 1706 | |||

Rsph3
Genomic Map