siRNA Details
| Gene Symbol: Pnldc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025724.1 | NP_001020895.1 | 5, 6 | 404 | ||
| XM_006227880.2 | XP_006227942.1 | 5, 6 | 1537 | |||
| XM_008758734.1 | XP_008756956.1 | 5, 6 | 1537 | |||
| XM_008758735.1 | XP_008756957.1 | 4, 5 | 707 | |||
| GenBank mRNA | BC099137.1 | - | 5, 6 | 404 | 7456056 | |

Pnldc1
Genomic Map