siRNA Details
| Gene Symbol: Eps8l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108467.1 | NP_001101937.1 | 16 | 1873 | ||
| XM_006228278.2 | XP_006228340.1 | 17 | 2152 | |||
| XM_006228279.2 | XP_006228341.1 | 17 | 2049 | |||
| XM_006228280.2 | XP_006228342.1 | 17 | 1924 | |||
| XM_006228282.2 | XP_006228344.1 | 17 | 2048 | |||
| XM_008758941.1 | XP_008757163.1 | 17 | 1945 | |||
| XM_008758942.1 | XP_008757164.1 | 17 | 1802 | |||
| XM_008758943.1 | XP_008757165.1 | 17 | 2152 | |||

Eps8l1
Genomic Map