siRNA Details
| Gene Symbol: Leng8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037790.2 | NP_001032879.2 | 14 | 2168 | ||
| XM_006228218.1 | XP_006228280.1 | 15 | 2393 | |||
| XM_006228219.2 | XP_006228281.1 | 15 | 2550 | |||
| XM_006228220.1 | XP_006228282.1 | 14 | 2282 | |||
| XM_008758946.1 | XP_008757168.1 | 15 | 2393 | |||
| XM_008758947.1 | XP_008757169.1 | 13 | 2231 | |||
| XM_008758948.1 | XP_008757170.1 | 14 | 2342 | |||

Leng8
Genomic Map