siRNA Details
| Gene Symbol: Ube2m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108471.1 | NP_001101941.1 | 5 | 952 | ||
| GenBank mRNA | FQ209516.1 | - | 6 | 545 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001243968, NM_145578, NM_001168469, BC026581, BC021792, and others.

Ube2m
Genomic Map