siRNA Details
| Gene Symbol: Vasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108475.2 | NP_001101945.2 | 12 | 1260 | ||
| XM_008758953.1 | XP_008757175.1 | 12 | 1839 | |||
| GenBank mRNA | BC168841.1 | - | 12 | 1260 | 9030574 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vasp| Gene Symbol: Vasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108475.2 | NP_001101945.2 | 12 | 1260 | ||
| XM_008758953.1 | XP_008757175.1 | 12 | 1839 | |||
| GenBank mRNA | BC168841.1 | - | 12 | 1260 | 9030574 | |