siRNA Details
| Gene Symbol: Zfp428 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108476.1 | NP_001101946.1 | 3 | 940 | ||
| XM_006228444.2 | XP_006228506.1 | 3 | 1030 | |||
| XM_006228445.2 | XP_006228507.1 | 3 | 1006 | |||
| XM_006228446.2 | XP_006228508.1 | 3 | 2329 | |||
| XM_006228447.2 | XP_006228509.1 | 3 | 2305 | |||
| XM_008758954.1 | XP_008757176.1 | 2 | 1683 | |||
| GenBank mRNA | FQ224206.1 | - | 3 | 635 | ||

Zfp428
Genomic Map