siRNA Details
| Gene Symbol: Atp5sl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009705.1 | NP_001009705.1 | 2, 3 | 216 | ||
| XM_006228449.2 | XP_006228511.1 | 2, 3 | 343 | |||
| GenBank mRNA | BC088283.1 | - | 2, 3 | 216 | 7384054 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp5sl| Gene Symbol: Atp5sl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009705.1 | NP_001009705.1 | 2, 3 | 216 | ||
| XM_006228449.2 | XP_006228511.1 | 2, 3 | 343 | |||
| GenBank mRNA | BC088283.1 | - | 2, 3 | 216 | 7384054 | |