siRNA Details
| Gene Symbol: Hnrnpul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108477.1 | NP_001101947.1 | 11 | 1762 | ||
| XM_008758955.1 | XP_008757177.1 | 11 | 1757 | |||
| XM_008758956.1 | XP_008757178.1 | 11 | 1659 | |||
| XM_008758957.1 | XP_008757179.1 | 11 | 1441 | |||
| GenBank mRNA | FQ222753.1 | - | 11 | 1775 | ||

Hnrnpul1
Genomic Map