siRNA Details
Gene Symbol: Sertad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007735.1 | NP_001007736.1 | 2 | 480 | ||
GenBank mRNA | BC061808.1 | - | 2 | 480 | 5598343 | |
FQ214111.1 | - | 2 | 494 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Sertad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007735.1 | NP_001007736.1 | 2 | 480 | ||
GenBank mRNA | BC061808.1 | - | 2 | 480 | 5598343 | |
FQ214111.1 | - | 2 | 494 |