siRNA Details
| Gene Symbol: Tyrobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212525.1 | NP_997690.1 | 2, 3 | 101 | ||
| GenBank mRNA | AY247021.1 | - | 2, 3 | 101 | ||
| FQ211365.1 | - | 2, 3 | 107 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tyrobp| Gene Symbol: Tyrobp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212525.1 | NP_997690.1 | 2, 3 | 101 | ||
| GenBank mRNA | AY247021.1 | - | 2, 3 | 101 | ||
| FQ211365.1 | - | 2, 3 | 107 | |||