siRNA Details
| Gene Symbol: Proser3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127579.1 | NP_001121051.1 | 9 | 986 | ||
| XM_006228772.1 | XP_006228834.1 | 9 | 1428 | |||
| XM_006228773.1 | XP_006228835.1 | 9 | 972 | |||
| XM_006228774.2 | XP_006228836.1 | 9 | 1081 | |||
| XM_008759167.1 | XP_008757389.1 | 8 | 1599 | |||
| XM_008759168.1 | XP_008757390.1 | 8 | 1596 | |||
| GenBank mRNA | BC166499.1 | - | 9 | 986 | 7114906 | |

Proser3
Genomic Map