siRNA Details
| Gene Symbol: Proser3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127579.1 | NP_001121051.1 | 2 | 117 | ||
| XM_006228772.1 | XP_006228834.1 | 2 | 559 | |||
| XM_006228773.1 | XP_006228835.1 | 2 | 103 | |||
| XM_006228774.2 | XP_006228836.1 | 2 | 212 | |||
| XM_008759167.1 | XP_008757389.1 | 1 | 730 | |||
| XM_008759168.1 | XP_008757390.1 | 1 | 730 | |||
| GenBank mRNA | BC166499.1 | - | 2 | 117 | 7114906 | |

Proser3
Genomic Map