siRNA Details
| Gene Symbol: Etv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228814.2 | XP_006228876.1 | 1 | 510 | ||
| XM_006228815.2 | XP_006228877.1 | 1 | 510 | |||
| XM_008759255.1 | XP_008757477.1 | 1 | 510 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Etv2| Gene Symbol: Etv2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006228814.2 | XP_006228876.1 | 1 | 510 | ||
| XM_006228815.2 | XP_006228877.1 | 1 | 510 | |||
| XM_008759255.1 | XP_008757477.1 | 1 | 510 | |||