siRNA Details
| Gene Symbol: Lgi4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199499.1 | NP_955793.1 | 2 | 422 | ||
| XM_008759169.1 | XP_008757391.1 | 2 | 937 | |||
| XM_008759170.1 | XP_008757392.1 | 2 | 136 | |||
| GenBank mRNA | BC064659.1 | - | 2 | 422 | 5623799 | |

Lgi4
Genomic Map