siRNA Details
| Gene Symbol: Ankrd27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271264.1 | NP_001258193.1 | 4 | 456 | ||
| XM_008759172.1 | XP_008757394.1 | 4 | 489 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankrd27| Gene Symbol: Ankrd27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271264.1 | NP_001258193.1 | 4 | 456 | ||
| XM_008759172.1 | XP_008757394.1 | 4 | 489 | |||