siRNA Details
| Gene Symbol: Vrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005561.1 | NP_001005561.1 | 12 | 1223 | ||
| XM_006229095.2 | XP_006229157.1 | 12 | 1282 | |||
| XM_006229096.2 | XP_006229158.1 | 12 | 1283 | |||
| GenBank mRNA | BC081928.1 | - | 12 | 1223 | 7113277 | |

Vrk3
Genomic Map