siRNA Details
| Gene Symbol: Aldh16a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033706.1 | NP_001028878.1 | 8 | 1013 | ||
| XM_006229099.2 | XP_006229161.1 | 8 | 1086 | |||
| XM_006229100.1 | XP_006229162.1 | 7 | 872 | |||
| GenBank mRNA | BC101860.1 | - | 8 | 1013 | 7936953 | |

Aldh16a1
Genomic Map