siRNA Details
| Gene Symbol: Snrnp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108483.1 | NP_001101953.1 | 7, 8 | 679 | ||
| XM_006229101.2 | XP_006229163.1 | 6, 7 | 623 | |||
| XM_006229102.2 | XP_006229164.1 | 6, 7 | 545 | |||
| GenBank mRNA | BC167051.1 | - | 6, 7 | 461 | 9089731 | |

Snrnp70
Genomic Map