siRNA Details
| Gene Symbol: Sytl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748924.3 | XP_003748972.1 | 7 | 1592 | ||
| XM_006229687.2 | XP_006229749.1 | 6 | 1460 | |||
| XM_006229694.2 | XP_006229756.1 | 7 | 1592 | |||
| XM_006229695.2 | XP_006229757.1 | 7 | 1593 | |||
| XM_008759651.1 | XP_008757873.1 | 7 | 1600 | |||
| XM_008759652.1 | XP_008757874.1 | 7 | 1751 | |||
| XM_008759653.1 | XP_008757875.1 | 7 | 4139 | |||
| XM_008759654.1 | XP_008757876.1 | 7 | 1600 | |||
| XM_008759655.1 | XP_008757877.1 | 7 | 1600 | |||
| XM_008759656.1 | XP_008757878.1 | 7 | 1600 | |||

Sytl2
Genomic Map