siRNA Details
| Gene Symbol: Sytl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748924.3 | XP_003748972.1 | 10 | 2093 | ||
| XM_006229687.2 | XP_006229749.1 | 9 | 5579 | |||
| XM_006229694.2 | XP_006229756.1 | 10 | 2093 | |||
| XM_006229695.2 | XP_006229757.1 | 10 | 2094 | |||
| XM_006229697.2 | XP_006229759.1 | 1 | 2335 | |||
| XM_008759651.1 | XP_008757873.1 | 10 | 5719 | |||
| XM_008759652.1 | XP_008757874.1 | 10 | 5870 | |||
| XM_008759653.1 | XP_008757875.1 | 10 | 8258 | |||
| XM_008759654.1 | XP_008757876.1 | 10 | 5719 | |||
| XM_008759656.1 | XP_008757878.1 | 10 | 5719 | |||

Sytl2
Genomic Map