siRNA Details
| Gene Symbol: Aamdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108493.1 | NP_001101963.1 | 5 | 762 | ||
| XM_006229768.1 | XP_006229830.1 | 4 | 671 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aamdc| Gene Symbol: Aamdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108493.1 | NP_001101963.1 | 5 | 762 | ||
| XM_006229768.1 | XP_006229830.1 | 4 | 671 | |||