siRNA Details
| Gene Symbol: P4ha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198775.1 | NP_942070.1 | 3 | 576 | ||
| XM_008759761.1 | XP_008757983.1 | 3 | 579 | |||
| XM_008759762.1 | XP_008757984.1 | 3 | 579 | |||
| GenBank mRNA | AY313450.1 | - | 3 | 576 | ||

P4ha3
Genomic Map