siRNA Details
| Gene Symbol: Fam168a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108494.1 | NP_001101964.1 | 5 | 508 | ||
| XM_006229773.2 | XP_006229835.1 | 4 | 1665 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam168a| Gene Symbol: Fam168a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108494.1 | NP_001101964.1 | 5 | 508 | ||
| XM_006229773.2 | XP_006229835.1 | 4 | 1665 | |||