siRNA Details
| Gene Symbol: Arap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229897.2 | XP_006229959.1 | 27 | 3921 | ||
| XM_006229898.2 | XP_006229960.1 | 27 | 4665 | |||
| XM_006229899.2 | XP_006229961.1 | 27 | 4663 | |||
| XM_006229900.1 | XP_006229962.1 | 25 | 3540 | |||
| XM_006229901.1 | XP_006229963.1 | 24 | 3361 | |||
| XM_008759810.1 | XP_008758032.1 | 28 | 5088 | |||
| XM_008759811.1 | XP_008758033.1 | 27 | 4663 | |||
| GenBank mRNA | BC088328.1 | - | 5 | 505 | ||

Arap1
Genomic Map