siRNA Details
| Gene Symbol: Olfml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013192.1 | NP_001013210.1 | 3 | 1105 | ||
| GenBank mRNA | BC081972.1 | - | 3 | 1105 | 7125978 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfml1| Gene Symbol: Olfml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013192.1 | NP_001013210.1 | 3 | 1105 | ||
| GenBank mRNA | BC081972.1 | - | 3 | 1105 | 7125978 | |