siRNA Details
| Gene Symbol: Akip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108497.1 | NP_001101967.1 | 3 | 572 | ||
| XM_006229987.1 | XP_006230049.1 | 4 | 461 | |||
| XM_006229988.2 | XP_006230050.1 | 4 | 392 | |||
| XM_006229989.1 | XP_006230051.1 | 4 | 431 | |||
| XM_006229990.1 | XP_006230052.1 | 4 | 461 | |||
| XM_006229991.1 | XP_006230053.1 | 3 | 380 | |||

Akip1
Genomic Map