siRNA Details
| Gene Symbol: Nrip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108498.1 | NP_001101968.1 | 1, 2 | 255 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nrip3| Gene Symbol: Nrip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108498.1 | NP_001101968.1 | 1, 2 | 255 | ||