siRNA Details
| Gene Symbol: Tead1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198589.1 | NP_001185518.1 | 2 | 445 | ||
| XM_006230052.2 | XP_006230114.1 | 2 | 727 | |||
| XM_006230053.2 | XP_006230115.1 | 2 | 727 | |||
| XM_006230054.2 | XP_006230116.1 | 2 | 727 | |||
| XM_006230055.2 | XP_006230117.1 | 2 | 727 | |||
| XM_006230056.2 | XP_006230118.1 | 2 | 727 | |||
| XM_006230057.2 | XP_006230119.1 | 2 | 727 | |||
| XM_006230058.2 | XP_006230120.1 | 2 | 727 | |||
| XM_006230059.2 | XP_006230121.1 | 2 | 1471 | |||
| XM_008759767.1 | XP_008757989.1 | 2 | 921 | |||
| XM_008759768.1 | XP_008757990.1 | 2 | 284 | |||
| GenBank mRNA | AY529194.1 | - | 1 | 175 | ||
| AY529195.1 | - | 1 | 175 | |||
| AY529196.1 | - | 1 | 175 | |||
| AY529197.1 | - | 1 | 175 | |||
| AY529198.1 | - | 1 | 175 | |||

Tead1
Genomic Map