siRNA Details
| Gene Symbol: Cyp2r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108499.1 | NP_001101969.1 | 5 | 803 | ||
| XM_006230081.2 | XP_006230143.1 | 5 | 1814 | |||
| XM_006230084.2 | XP_006230146.1 | 5 | 1593 | |||
| GenBank mRNA | BC168986.1 | - | 4 | 1221 | ||
| FQ219437.1 | - | 4 | 1117 | |||

Cyp2r1
Genomic Map