siRNA Details
| Gene Symbol: LOC361635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017463.1 | NP_001017463.1 | 8 | 724 | ||
| XM_006230122.2 | XP_006230184.1 | 8 | 841 | |||
| GenBank mRNA | BC083805.1 | - | 8 | 724 | 7109216 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC361635| Gene Symbol: LOC361635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017463.1 | NP_001017463.1 | 8 | 724 | ||
| XM_006230122.2 | XP_006230184.1 | 8 | 841 | |||
| GenBank mRNA | BC083805.1 | - | 8 | 724 | 7109216 | |