siRNA Details
| Gene Symbol: Acsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014162.4 | NP_001014184.3 | 13 | 1875 | ||
| XM_006230123.1 | XP_006230185.1 | 14 | 1818 | |||
| XM_006230126.1 | XP_006230188.1 | 14 | 1818 | |||
| GenBank mRNA | AY325244.1 | - | 2207 | |||
| BC078915.1 | - | 13 | 1874 | 7096091 | ||

Acsm5
Genomic Map